Anti-HOXA1 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
28-188
PRSI28-188EA 601 EUR InStock
PRSI28-188
Anti-HOXA1 Rabbit Polyclonal Antibody
Anticorps
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA1 is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development.
Antibody isMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 µg 100 µG PRSI28-188
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité