Anti-ECT2 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
30-812
PRSI30-812EA 660 EUR InStock
PRSI30-812
Anti-ECT2 Rabbit Polyclonal Antibody
Anticorps
ECT2 is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of ECT2 is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.Western blots using two different antibodies against two unique regions of thisMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
50 µg 50 µG PRSI30-812
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité