Anti-TRAF6 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
3129
PRSI3129EA 435 EUR InStock
PRSI3129
Anti-TRAF6 Rabbit Polyclonal Antibody
Anticorps
TRAF6 Antibody: Signals from the IL-1 receptor (IL-1R)/Toll-like receptor (TLR) and TNF receptor (TNFR) superfamilies are critical for regulating the function of antigen-presenting cells. Signals transduced by these molecules lead to increased expression and activation of transcription factors such as NF-kappa B. TNF receptor-associated factor 6 (TRAF6) is unique in that it is a signaling adapter molecule common to both families. TRAF6 is important in cytokine production, dendritic cell (DC) maturation, and the T cell stimulatory capacity of DCs in response to TLR and CD40 ligands. It can beMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité