Anti-CDC20 Mouse Monoclonal Antibody [clone: AR12]
Fournisseur: ProSci Inc.
33-106
PRSI33-106EA 773 EUR InStock
PRSI33-106
Anti-CDC20 Mouse Monoclonal Antibody [clone: AR12]
Anticorps
Cyclins, regulatory subunits which associate with kinases, control many of the important steps in cell cycle progression. The Cdc2 kinase (p34Cdc2) exhibits kinase activity in vitro and exists in a complex with both cyclin B and a protein homologous to p13SUC1. Cdc2 kinase is the active subunit of the M phase promoting factor (MPF) and the M phase-specific Histone H1 kinase. The p34Cdc2/cyclin B complex is required for the G2 to M transition. An additional cell cycle-dependent protein kinase, termed p55cdc, exhibits a high degree of homology with the S. cerevisiae proteins Cdc20 and Cdc4.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité