Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-HSPD1 Mouse Monoclonal Antibody [clone: LK1]

Fournisseur: ProSci Inc.
33-141
PRSI33-141EA 773 EUR InStock
PRSI33-141
Anti-HSPD1 Mouse Monoclonal Antibody [clone: LK1]
Anticorps
This antibody recognises a 60 kDa protein, identified as the heat shock protein 60. The antibody's epitope is localized between amino acids 383-447 of human HSP60. A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp’s), more appropriately called as stress response proteins (srp’s). HSP60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial HSP60, but also against its mammalian homolog. Clone LK1 antibody, unlike LK2 antibody, recognises only the mammalian (not bacterial) HSP60 and is useful in distinguishing HSP60 from mammals and bacteria.

Flow Cytometry: 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml WB: 0,25-0,5 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be titered up or down for optimal performance.

1. Staining of formalin-fixed tissues is enhanced by boiling tissue sections in 10 mM Citrate Buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: LK1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse, Rat, Hamster, Sheep, Rabbit, Bovine, Dog, Pig, Monkey, Chicken, Xenopus, Drosophila

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité