Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-IgG Fc Mouse Monoclonal Antibody [clone: IG266]

Fournisseur: ProSci Inc.
33-145
PRSI33-145EA 773 EUR InStock
PRSI33-145
Anti-IgG Fc Mouse Monoclonal Antibody [clone: IG266]
Anticorps

Immunoglobulin gamma (IgG) is the most common class of antibody in blood and extracellular fluid. Approximately 75% of serum antibodies in humans are IgG. There are four immunoglobulin gamma subclasses: one, two, three and four. IgG1 is the most common, with 68% of all gamma class antibodies being G1, and G4 is the least common at 4%. Gamma class antibodies are found primarily in the secondary immune response, class switching from IgM and IgD. They are the only class of antibody that can cross the placenta, and along with IgA secreted in breast milk, provide the neonate with humoral immunity before immune system development occurs.

This antibody recognises a protein of 75 kDa identified as the gamma heavy chain of human immunoglobulins. It does not cross-react with alpha, mu, epsilon, or delta heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. The IgG antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.


Flow Cytometry: 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the anti-IgG antibody to be titered up or down for optimal performance.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: IgG Fc
Clonality: Monoclonal
Clone: IG266
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité