Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-PMEL Mouse Monoclonal Antibody [clone: HMB45]

Fournisseur: ProSci Inc.
33-201
PRSI33-201EA 773 EUR InStock
PRSI33-201
Anti-PMEL Mouse Monoclonal Antibody [clone: HMB45]
Anticorps
Melanocytes produce organelles called melanosomes which produce melanin, the pigment that gives color to skin, hair, eyes, scales and feathers. gp100 was identified in an attempt to clone the gene Tyrosinase, an enzyme required for melanin synthesis. Further testing determined that gp100 is a melanoma-specific protein and is responsible for melanosome maturation, facilitating the transition from amorphous rounded vesicles to fibrillary ellipsoid organelles.
Metastatic amelanotic melanoma can often be confused with a variety of poorly differentiated carcinomas, large cell lymphomas, and sarcomas using H & E stains alone. It is also difficult to differentiate melanoma from spindle cell carcinomas and various types of mesenchymal neoplasms. Clone HMB45 gp100 antibody stains fetal and neonatal melanocytes, junctional and blue nevus cells, and malignant melanoma. It also stains angiomyolipomas, tumors most commonly associated with the kidney. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. This gp100 antibody does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin.

Western Blot: 1 - 2 µg/ml Flow Cytometry: 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml IHC (FFPE): 0,5-1,0 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the gp100 antibody to be titered up or down for optimal performance.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: PMEL
Clonality: Monoclonal
Clone: HMB45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité