Anti-CD19 Mouse Monoclonal Antibody [clone: CVID3/429]
Fournisseur: ProSci Inc.
33-284
PRSI33-284EA 773 EUR InStock
PRSI33-284
Anti-CD19 Mouse Monoclonal Antibody [clone: CVID3/429]
Anticorps
CD19 is expressed on follicular dendritic cells and B cells. It is present on B cells from earliest recognizable B-lineage cells during development to B-cell blasts but is lost on maturation to plasma cells. It primarily acts as a B cell co-receptor in conjunction with CD21 and CD81. Upon activation, the cytoplasmic tail of CD19 becomes phosphorylated, which leads to binding by Src-family kinases and recruitment of PI-3 kinase. [Wiki].
Flow Cytometry: 0,5-1 µg/million cells Immunofluorescence: 0,5 - 1 µg/ml Variations in protocols, secondaries and substrates may require the CD19 antibodyMore Product Information
New Product
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 µg 100 µG PRSI33-284
New Product
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité