Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-TP53 Mouse Monoclonal Antibody [clone: PAb 122]

Fournisseur: ProSci Inc.
33-854
PRSI33-854EA 773 EUR InStock
PRSI33-854
Anti-TP53 Mouse Monoclonal Antibody [clone: PAb 122]
Anticorps
This antibody is specific to a conserved determinant of the p53 protein. PAb 122 binds to the C-terminus (aa 370-378) of both wild type and mutated p53. When microinjected into nuclei, PAb 122 blocked re-entry into the S-phase of the cell cycle. Mutation and/or allelic loss of p53 is one of the causes of a variety of mesenchymal and epithelial tumors. If it occurs in the germ line, such tumors run in families. p53 binds to a DNA consensus sequence, the p53 response element, and it regulates normal cell growth cycle events by activating transcription of genes, involved either in progression through the cycle, or causing arrest in G1 when the genome is damaged. In most transformed and tumor cells the concentration of p53 is increased 5-1000 fold over the minute concentrations (1000 molecules cell) in normal cells, principally due to the increased half-life (4 h) compared to that of the wild-type (20 min). The protein in the nucleus, but is detectable at the plasma membrane during mitosis and when certain mutations modulate cytoplasmic/nuclear distribution. Its the most commonly mutated gene in spontaneously occurring human cancers. Mutations arise with an average frequency of 70% but incidence varies from zero in carcinoid lung tumors to 97% in primary melanomas. High concentrations of p53 protein are transiently expressed in human epidermis and superficial dermal fibroblasts following mild ultraviolet irradiation.

Western blot: 0,5 - 1 µg/ml Flow Cytometry: 0,5-1 µg/million cells Immunofluorescence: 0,5 - 1 µg/ml IHC (Frozen) only: 0,5 - 1 µg/ml for 30 min at RT Variations in protocols, secondaries and substrates may require the p53 antibody to be titered up or down for optimal performance.

Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: PAb 122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human, Monkey, Dog, Mouse, Rat, Hamster

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité