Anti-NCAM1 Mouse Monoclonal Antibody [clone: CDLA56]
Fournisseur: ProSci Inc.
33-982
PRSI33-982EA 773 EUR InStock
PRSI33-982
Anti-NCAM1 Mouse Monoclonal Antibody [clone: CDLA56]
Anticorps
CD56/NCAM is a cell adhesion protein which is a member of the immunoglobulin superfamily. It is involved in cell-to-cell interactions as well as cell-matrix interactions during development and differentiation. CD56 has been shown to be involved in development of the nervous system, and for cells involved in the expansion of T cells and dendritic cells which play an important role in immune surveillance. [UniProt].
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1)More Product Information
New Product
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 µg 100 µG PRSI33-982
New Product
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité