Anti-CEACAM5 Mouse Monoclonal Antibody [clone: SPM541]
Fournisseur: ProSci Inc.
34-093
PRSI34-093EA 0 EUR InStock
PRSI34-093
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: SPM541]
Anticorps
This antibody recognises proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesised during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This mAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci ofMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité