Anti-MUC2 Mouse Monoclonal Antibody [clone: SPM296]
Fournisseur: ProSci Inc.
34-135
PRSI34-135EA 773 EUR InStock
PRSI34-135
Anti-MUC2 Mouse Monoclonal Antibody [clone: SPM296]
Anticorps
Recognises a single glycoprotein of 520 kDa, identified as mucin 2 (MUC2). This mAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3. Mucins are high molecular weight glycoproteins, which constitute the major component of the mucus layer that protects the gastric epithelium. MUC2 is specifically expressed in goblet cells of the small intestine & colon; in about 65% of colonic carcinomas and about 40% of gastric carcinomas. MUC2 is rarely expressed outside of the GI tract with the exceptions of mucinous carcinoma of breast and clear cell-type carcinomas of theMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité