Anti-Verotoxins Mouse Monoclonal Antibody
Fournisseur: ProSci Inc.
35-672
PRSI35-672EA 588 EUR InStock
PRSI35-672
Anti-Verotoxins Mouse Monoclonal Antibody
Anticorps
Verotoxins (Shiga-like) antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.
Verotoxins antibody (Shiga-like) is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Verotoxins
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity: Bacteria
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité