Anti-CTRP2 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
3561
PRSI3561EA 435 EUR InStock
PRSI3561
Anti-CTRP2 Rabbit Polyclonal Antibody
Anticorps
CTRP2 Antibody: Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscleMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 µg 100 µG PRSI3561
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité