Human recombinant IL17A (from E. coli)
Fournisseur: ProSci Inc.
40-521
PRSI40-521EA 412 EUR InStock
PRSI40-521
Human recombinant IL17A (from E. coli)
Protéines et Peptides
Human IL-17A is a homodimer of two 136 amino acid chains, secreted by activated T-cells that act on stromal cells to induce production of proinflammatory and hematopoietic bioactive molecules. Today, IL-17 represents a family of structurally-related cytokines that share a highly conserved C-terminal region but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17A exhibits cross-species bioactivity between human and murine cells. Recombinant human IL-17A is a 31.0 kDa disulfide-linked homodimer of two 136 amino acid polypeptide chains.
  • Murine IL-17A is a disulfide linked homodimer, secreted by activated T-cells that act on stromal cells to induce production of proinflammatory and hematopoietic bioactive molecules. Today, IL-17 represents a family of structurally-related cytokines that share a highly conserved C-terminal region but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17A exhibits cross-species bioactivity between human and murine cells. Recombinant murine IL-17A is a 30.0 kDa disulfide-linked homodimer of two 133 amino acid polypeptide chains.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité