Human recombinant MCP1 (from E. coli)
Fournisseur: ProSci Inc.
40-564
PRSI40-564EA 412 EUR InStock
PRSI40-564
Human recombinant MCP1 (from E. coli)
Protéines et Peptides
The MCP proteins belong to the CC chemokine family, and signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross reacts across species. Recombinant human MCP-1 is an 8.6 kDa protein containing 76 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité