Human Recombinant NT3 (from E. coli)
Fournisseur: ProSci Inc.
40-565
PRSI40-565EA 461 EUR InStock
PRSI40-565
Human Recombinant NT3 (from E. coli)
Protéines et Peptides
NT-3 is a neurotrophic factor structurally related to β-NGF, BDNF, and NT-4. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. NT-3 is expressed by neurons of the central nervous systems and can signal through the trk receptors. NT-3 promotes the growth and survival of nerve and glial cells. The amino acid sequences of human, murine and rat NT-3 are identical. Recombinant human NT-3 is a noncovalently linked homodimer, of two 13.6 kDa polypeptide monomers (240 total amino acid residues). Human and Mouse NT-3 sequences are identical.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité