Human Recombinant NGF beta (from E. coli)
Fournisseur: ProSci Inc.
40-572
PRSI40-572EA 461 EUR InStock
PRSI40-572
Human Recombinant NGF beta (from E. coli)
Protéines et Peptides
Beta-NGF is a neurotrophic factor structurally related to BDNF, NT-3 and NT-4. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. Beta-NGF is a potent neurotrophic factor that signals through its receptor Beta-NGFR, and plays a crucial role in the development and preservation of the sensory and sympathetic nervous systems. Beta-NGF also acts as a growth and differentiation factor for B lymphocytes and enhances B-cell survival. The functional form of human Beta-NGF is a noncovalently disulfide-linked homodimer, of two 13.5 kDa polypeptide monomers (238 total amino acid residues). The three disulfide bonds are required for biological activity.Manufactured using all non-animal reagents.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité