Human recombinant FGF10 (from E. coli)
Fournisseur: ProSci Inc.
40-587
PRSI40-587EA 461 EUR InStock
PRSI40-587
Human recombinant FGF10 (from E. coli)
Protéines et Peptides
FGF-10 is a heparin binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-10 is most related to KGF/FGF-7 and is expressed during the development and preferentially in adult lungs. It signals through the FGFR 2b. Recombinant human FGF-10 is a 19.3 kDa protein consisting of 170 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité