Human recombinant FGF Acidic (from E. coli)
Fournisseur: ProSci Inc.
40-588
PRSI40-588EA 412 EUR InStock
PRSI40-588
Human recombinant FGF Acidic (from E. coli)
Protéines et Peptides
FGF-acidic is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-acidic is a non-glycosylated heparin binding growth factor that is expressed in the brain, kidney, retina, smooth muscle cells, bone matrix, osteoblasts, astrocytes and endothelial cells. FGF-acidic has the ability to signal through all the FGF receptors. Recombinant murine FGF-acidic is a 15.9 kDa protein consisting of 141 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité