Human recombinant IL2 (fromE. coli)
Fournisseur: ProSci Inc.
40-599
PRSI40-599EA 461 EUR InStock
PRSI40-599
Human recombinant IL2 (fromE. coli)
Protéines et Peptides
IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions which are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine activated killer cells, monocytes, macrophages and oligodendrocytes. Recombinant murine IL-2 is a 17.2 kDa protein, containing 149 amino acid residues. Recombinant rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues. Recombinant human IL-2 is a 15.5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité