Human recombinant IL4 (from E. coli)
Fournisseur: ProSci Inc.
40-604
PRSI40-604EA 461 EUR InStock
PRSI40-604
Human recombinant IL4 (from E. coli)
Protéines et Peptides
IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival and gene expression. Produced by mast cells, T cells and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells, characterized by their cytokine-secretion profile that includes secretion of IL-4, IL-5, IL-6, IL-10, and IL-13, which favor a humoral immune response. Another dominant function of IL-4 is the regulation of immunoglobulin class switching to the IgG1 and IgE isotypes. Excessive IL-4 production by Th2 cells has been associated with elevated IgE production and allergy. Recombinant human IL-4 is a 14.9 kDa globular protein containing 129 amino acid residues. Recombinant rat IL-4 is a 14.0 kDa globular protein containing 126 amino acid residues. Recombinant murine IL-4 is a 13.5 kDa globular protein containing 120 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité