Human recombinant KGF (from E. coli)
Fournisseur: ProSci Inc.
40-605
PRSI40-605EA 461 EUR InStock
PRSI40-605
Human recombinant KGF (from E. coli)
Protéines et Peptides
Keratinocyte Growth Factor (KGF/FGF-7) is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of variety of tissues, by promoting cellular proliferation and differentiation. KGF/FG-7 is a mitogen factor specific for epithelial cells and keratinocytes and signals through FGFR 2b. KGF/FGF-7 plays a role in kidney and lung development, angiogenesis, and wound healing. Recombinant human KGF/FGF-7 is an 18.9 kDa protein consisting of 163 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité