Human recombinant VEGF121 (from E. coli)
Fournisseur: ProSci Inc.
40-612
PRSI40-612EA 461 EUR InStock
PRSI40-612
Human recombinant VEGF121 (from E. coli)
Protéines et Peptides
VEGF is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. VEGF signals through three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Due to its increased acidity, VEGF121 circulates more freely than other VEGF forms, which bind more tightly with vascular heparin sulfates. Recombinant human VEGF121 is a 28.4 kDa disulfide-linked homodimeric protein consisting of two 121 amino acid polypeptide chains.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité