Human recombinant VEGF165 (from E. coli)
Fournisseur: ProSci Inc.
40-613
PRSI40-613EA 461 EUR InStock
PRSI40-613
Human recombinant VEGF165 (from E. coli)
Protéines et Peptides
VEGF is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. Substantial evidence implicates VEGF in the induction of tumor metastasis and intra-ocular neovascular syndromes. VEGF signals through the three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant human VEGF is a 38.2 kDa disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains. Recombinant rat VEGF is a 38.5 kDa disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité