Mouse recombinant IL3 (from E. coli)
Fournisseur: ProSci Inc.
40-618
PRSI40-618EA 461 EUR InStock
PRSI40-618
Mouse recombinant IL3 (from E. coli)
Protéines et Peptides
IL-3 is a hematopoietic growth factor that promotes the survival, differentiation and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil and mast cell lineages. Produced by T cells, mast cells and eosinophils, IL-3 enhances thrombopoieses, phagocytosis, and antibody-mediated cellular cytotoxicity. Its ability to activate monocytes suggests that IL-3 may have additional immunoregulatory roles. Many of the IL-3 activities depend upon co-stimulation with other cytokines. IL-3 is species-specific, variably glycosylated cytokine. Recombinant murine IL-3 is a 15.1 kDa globular protein containing 135 amino acid residues. Recombinant human IL-3 is a 15.0 kDa globular protein containing 133 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité