Anti-FABP7 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
4259
PRSI4259EA 435 EUR InStock
PRSI4259
Anti-FABP7 Rabbit Polyclonal Antibody
Anticorps
FABP7 Antibody: FABP7 was initially isolated from a human fetal brain cDNA library and whose mRNA was expressed in adult brain and muscle tissues at low levels. The protein encoded by this gene is a member of the fatty acid binding protein (FABPs) family, a group of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs are thought to play roles in fatty acid uptake, transport, and metabolism. FABP7 is a downstream gene of the Pax6 transcription factor and has been suggested to be essential for the maintenance of neuroepithelialMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 µg 100 µG PRSI4259
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité