Human Synthetic CD38 Peptide
Fournisseur: ProSci Inc.
43-487P
PRSI43-487PEA 357 EUR InStock
PRSI43-487P
Human Synthetic CD38 Peptide
Protéines et Peptides
CD38 peptide is used for blocking the activity of CD38 antibody (catalog number 43-487 ).
The CD38 antibody ships at 0,5 mg/ml and the STK36 Peptide ships as a 100 µg pellet. When peptide is reconstituted in 200 µL water, the concentration will be 0,5 mg/ml, identical to that of the antibody. The best starting ratio would be 1:1 (which means molar excess of peptide relative to antibody when identical volumes are mixed). Mix equal volume of peptide and antibody at the required dilution and leave at ambient temperature for 20 minutes. Then incubate and develop in parallel two blots: one with the antibody and one with the antibody which has been pre-absorbed with the provided peptide.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité