Anti-ITIH5 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
55-530
PRSI55-530EA 620 EUR InStock
PRSI55-530
Anti-ITIH5 Rabbit Polyclonal Antibody
Anticorps
This gene encodes a heavy chain component of one of the inter-alpha-trypsin inhibitor (ITI) family members. ITI proteins are involved in extracellular matrix stabilization and in the prevention of tumor metastasis. They are also structurally related plasma serine protease inhibitors and are composed of a light chain and varying numbers of heavy chains. This family member is thought to function as a tumor suppressor in breast and thyroid cancers. Alternative splicing results in multiple transcript variants.
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité