Anti-BTLA Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
7473
PRSI7473EA 435 EUR InStock
PRSI7473
Anti-BTLA Rabbit Polyclonal Antibody
Anticorps
The B- and T-lymphocyte attenuator (BTLA) protein is a member of the immunoglobulin superfamily containing a single immunoglobulin (Ig) domain. Like other coihibitory receptors such as CTLA-4 and PD-1, BTLA functions as a receptor that relays inhibitory signals to suppress the immune response, and can inhibit LPS-induced endotoxic shock by suppressing TLR4 signaling in innate immune cells (1,2). Polymorphisms in this gene have also been associated with an increased risk of rheumatoid arthritis.
BTLA Antibody is affinity chromatography purified via peptide column.
BTLA Antibody isMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité