Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (FITC (Fluorescein Isothiocyanate))
Fournisseur: ProSci Inc.
76-323
PRSI76-323EA 224 EUR InStock
PRSI76-323
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (FITC (Fluorescein Isothiocyanate))
Anticorps
The OKT4 monoclonal antibody specifically binds to the CD4 receptor for the human immunodeficiency virus (HIV). CD4 is a 59 kDa single-chain transmembrane glycoprotein that expressed on the surface of most of the thymocytes, T-helper cells, and in low levels on monocytes and macrophages. CD4 is a co-receptor in the antigen-induced T cell activation (together with the MHC class II). The OKT4 and the RPA-T4 monoclonal antibodies recognize different epitopes of CD4 and they do not exhibit cross-block binding.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: FITC (More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité