Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (VioletFluor® 450)
Fournisseur: ProSci Inc.
76-354
PRSI76-354EA 0 EUR InStock
PRSI76-354
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (VioletFluor® 450)
Anticorps
The RPA-T4 monoclonal antibody specifically binds to the CD4 receptor for the human immunodeficiency virus (HIV). CD4 is a 59 kDa single-chain transmembrane glycoprotein that expressed on the surface of most of the thymocytes, T-helper cells, and in low levels on monocytes and macrophages. CD4 is a co-receptor in the antigen-induced T cell activation, together with the MHC class II. The RPA-T4 antibody is capable of blocking HIV binding and inhibiting syncytium formation, by binding to the D1 domain of the CD4 antigen. The OKT4 and the RPA-T4 monoclonal antibodies recognize differentMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité