Anti-Cd19 Rat Monoclonal Antibody [clone: 1D3] (FITC (Fluorescein Isothiocyanate))
Fournisseur: ProSci Inc.
76-778
PRSI76-778EA 224 EUR InStock
PRSI76-778
Anti-Cd19 Rat Monoclonal Antibody [clone: 1D3] (FITC (Fluorescein Isothiocyanate))
Anticorps
The 1D3 monoclonal antibody specifically reacts with mouse CD19, a 95 kDa transmembrane glycoprotein, a member of the Ig superfamily and a B cell-lineage differentiation antigen expressed by all the B lymphocyte development stages, except for the terminally differentiated plasma cells. CD19 associates with CD21, CD81 and MHC class II to form a multi-molecular complex that initiates the mature B lymphocyte activation by interaction with the B-cell receptors. CD 19 enhances the B cell proliferation, development and activation, and the maturation of memory B cells. In CD19-deficient mice, theMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité