Anti-ICOS Mouse Monoclonal Antibody [clone: ISA-3] (PE (Phycoerythrin)-Cy7®)
Fournisseur: ProSci Inc.
76-969
PRSI76-969EA 628 EUR InStock
PRSI76-969
Anti-ICOS Mouse Monoclonal Antibody [clone: ISA-3] (PE (Phycoerythrin)-Cy7®)
Anticorps
The ISA-3 monoclonal antibody specifically reacts with human CD278 or Inducible COStimulatory molecule (ICOS), a 47 to 60 kDa homodimeric glycoprotein also known as H4, AILIM, and CRP-1. It is expressed on activated T cells and a group of thymocytes and is involved in the proliferation, activation, and cytokine production of T cells. When plate-bound, the ISA-3 antibody induces the production of IL-4,5,10 in T cells.
Type: Primary
Antigen: ICOS
Clonality: Monoclonal
Clone: ISA-3
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité