Anti-HTNVsSgp1 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
7689
PRSI7689EA 435 EUR InStock
PRSI7689
Anti-HTNVsSgp1 Rabbit Polyclonal Antibody
Anticorps
Hantaan virus (HNTV) is the prototype virus of the genus hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA virus that is the eiological agent of Korean hemorrhagic fever. Infection of cultured Vero E6 cells with HNTV induced expression of the heat shock protein HSP70. A substantial fraction of HSP70 was found to be associated with the HNTV nucleocapsid (NP). Vero E6 cells that overexpress HSP70 show significantly reduced levels of NP and an increased resistance to infection by HNTV.
Hantaan virus nucleocapsid antibody is affinity chromatography purified via peptideMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité