Anti-Klrb1c Mouse Monoclonal Antibody [clone: PK136] (FITC (Fluorescein Isothiocyanate))
Fournisseur: ProSci Inc.
77-159
PRSI77-159EA 232 EUR InStock
PRSI77-159
Anti-Klrb1c Mouse Monoclonal Antibody [clone: PK136] (FITC (Fluorescein Isothiocyanate))
Anticorps
The PK136 monoclonal antibody is specific for the mouse NK1.1, a receptor from the killer cell lectin-like receptor (KLR) family. Nk1.1 is an antigen encoded by the Klrb1c/NKR-P1C gene expressed by the natural killer cells of some selected strains of mice (C57BL, FVB/N, NZB) and encoded by the Klrb1b/NKR-P1B gene expressed on Swiss NIH and SJL mice. Pk136 binds to an epitope common to NKR-P1B and NKR-P1C.The Klrb1 is a family of type II transmembrane C-type lectin receptors. Klrb1c activates the NK-cell cytotoxicity, while Klrb1b inhibits it. PK136 is useful in defining the NK cells and theMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité