Mouse recombinant IDO (from E. coli)
Fournisseur: ProSci Inc.
90-017
PRSI90-017EA 813 EUR InStock
PRSI90-017
Mouse recombinant IDO (from E. coli)
Protéines et Peptides
IDO catalyses the first and rate-limiting step in the main pathway of human tryptophan catabolism, the kynurenine pathway. Proinflammatory mediators, such as endotoxin and IFN-gamma induce the expression of IDO in several tissues. IDO-dependent suppression of T cell responses might function as natural immunoregulatory mechanism. Physiological IDO activity has been implicated in T cell tolerance to tumours, dysfunctional selftolerance in non-obese diabetic (NOD) mice, and as a protective negative regulator in autoimmune disorders.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité