Human recombinant FABP4 (from E. coli)
Fournisseur: ProSci Inc.
90-023
PRSI90-023EA 488 EUR InStock
PRSI90-023
Human recombinant FABP4 (from E. coli)
Protéines et Peptides
FABP4 is a fatty acid binding protein in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs regulate the fatty acid uptake, transport, and metabolism. aP2, the mouse form of FABP4, seems to be central to the pathway that links obesity to insulin resistance. aP2 was shown to be an adipokine linking adipocytes to hepatic glucose production and that neutralising secreted aP2 may represent an effective therapeutic strategy against diabetes.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité