Human Recombinant GPX3 (from HEK293 cells)
Fournisseur: ProSci Inc.
90-026
PRSI90-026EA 930 EUR InStock
PRSI90-026
Human Recombinant GPX3 (from HEK293 cells)
Protéines et Peptides
Glutathione peroxidase 3 (GPX3) is a selenium-dependent extracellular enzyme, protecting cells and enzymes from oxidative damage by catalysing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxides. GPX3 is involved in inflammatory bowel disease, in asthma and in diabetes. The activity of GPX3 is significantly reduced in the plasma and tissue of cancer patients. Recently, GPX3 was suggested to serve as a molecular marker for the diagnosis of clear cell type ovarian adenocarcinoma.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité