Rat Recombinant RBP4 (from HEK293 cells)
Fournisseur: ProSci Inc.
90-041
PRSI90-041EA 365 EUR InStock
PRSI90-041
Rat Recombinant RBP4 (from HEK293 cells)
Protéines et Peptides
Retinol binding protein 4 (RBP4; RBP) is a 21kDa secreted protein, a member of the lipocalin family and is known as the primary transporter of retinol (vitamin A) to tissues. A recent report revealed RBP4 as an adipokine linking glucose transporter 4 (GLUT4) suppression in adipose tissue to insulin. Elevated human and mouse serum RBP4 levels are associated with insulin resistance and its severity, obesity, and certain components of metabolic syndrome. Furthermore, human serum RBP4 levels are closely related to renal function.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité