Human recombinant ST2 (from HEK293 cells)
Fournisseur: ProSci Inc.
90-054
PRSI90-054EA 813 EUR InStock
PRSI90-054
Human recombinant ST2 (from HEK293 cells)
Protéines et Peptides
The ST2 (Interleukin-1 receptor-like 1; Interleukin-33 receptor) gene was originally identified as a gene induced by serum or oncogene expression in fibroblasts. The gene produces a shorter soluble secreted form (ST2) and a longer, transmembrane form (ST2L) by alternative splicing. Soluble ST2 has been shown to downregulate the expression of TLR1 and TLR4. ST2L negatively regulates TLR4 signalling and induces endotoxin tolerance, and enhances Th2 responses. IL-33 is the specific ligand for ST2L.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité