Human recombinant MFAP4 (from HEK293 cells)
Fournisseur: ProSci Inc.
90-069
PRSI90-069EA 852 EUR InStock
PRSI90-069
Human recombinant MFAP4 (from HEK293 cells)
Protéines et Peptides
Microfibril-associated protein 4 (MFAP4) was initially identified as a gene commonly deleted in the contiguous Smith-Magenis syndrome (SMS) gene. MFAP4 was identified as growth factor to support expansion of hematopoietic stem cells in culture. It is involved in calcium-dependent cell adhesion and intercellular interactions.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité