Human recombinant Calreticulin (from E. coli)
Fournisseur: ProSci Inc.
90-137
PRSI90-137EA 448 EUR InStock
PRSI90-137
Human recombinant Calreticulin (from E. coli)
Protéines et Peptides
Calreticulin is involved in regulation of intracellular Ca2+ homoeostasis and ER Ca2+ capacity. It constitutes together with calnexin and ERp57 the ‘calreticulin/calnexin cycle’ that is responsible for folding and quality control of newly synthesised glycoproteins. Calreticulin has been implicated to play a role in many biological systems, including functions inside and outside the ER, indicating that the protein is a multi-process molecule. Recently, Calreticulin was shown to enhance the merger of macrophages and tumour cells, increasing phagocytosis.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité