Human recombinant Jagged-2 (from HEK293 cells)
Fournisseur: ProSci Inc.
90-167
PRSI90-167EA 488 EUR InStock
PRSI90-167
Human recombinant Jagged-2 (from HEK293 cells)
Protéines et Peptides
Jagged-2 is a putative Notch ligand involved in the mediation of Notch signalling, that can induce stromal cells to secrete IL-6, VEGF and IGF-1. Notch activation can interact with NF-kappaB and C-myc to promote the proliferation and to inhibit the apoptosis of MM cells, showing in the relationship between the incidence of myeloma and drug resistance. Jagged-2 is also involved in limb development.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité