Human recombinant Contactin-6 (from HEK293 cells)
Fournisseur: ProSci Inc.
90-169
PRSI90-169EA 852 EUR InStock
PRSI90-169
Human recombinant Contactin-6 (from HEK293 cells)
Protéines et Peptides
Contactin-6 is a member of the contactins, which mediate cell surface interactions during nervous system development. Contactin-6 has a key role in the formation of glutamatergic, but not GABAergic, synapses during postnatal development of the hippocampal formation as well as the cerebellum. It is involved in oligodendrocytes generation by acting as a ligand of Notch1. It is also involved in motor coordination.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité