Human Recombinant HES1 (from E. coli)
Fournisseur: ProSci Inc.
90-193
PRSI90-193EA 1190 EUR InStock
PRSI90-193
Human Recombinant HES1 (from E. coli)
Protéines et Peptides
bHLH proteins are essential for neurogenesis, myogenesis, haematopoiesis and sex determination. HES1 (Hairy and Enhancer of Split 1) is a basic Helix-Loop-Helix (bHLH) transcriptional repressor regulated by the Notch pathway and expressed in various tissues. HES1 is well known in development of different tissues, particularly endoderm derived endocrine organs like pancreas and neuroendocrine cells of lung. HES1 controls normal development by regulating proliferation and differentiation and so cell fate. Notch signalling plays both oncogenic and tumour suppressor roles depending on cell type. In contrast to T cell acute lymphoblastic leukaemia (ALL), where Notch activation promotes leukemogenesis, induction of Notch signalling in B cell ALL (B-ALL) leads to growth arrest and apoptosis. The Notch target HES1 is sufficient to reproduce this tumour suppressor phenotype in B-ALL by interacting and activating PARP-1 and regulating proapoptotic signals.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité