Human recombinant Annexin V (from E. coli), ATTO 647N
Fournisseur: ProSci Inc.
90-220
PRSI90-220EA 409 EUR InStock
PRSI90-220
Human recombinant Annexin V (from E. coli), ATTO 647N
Protéines et Peptides
Annexin V is an anticoagulant protein that acts as an indirect inhibitor of the thromboplastin-specific complex, which is involved in the blood coagulation cascade. Annexin V competes for phosphatidylserine binding sites (e. g. platelets) with prothrombin or coagulation factors and inhibits the activity of phospholipase A1. Antibodies directed against annexin V are found in patients with a disease called the antiphospholipid syndrome(APS), a thrombophilic disease associated with autoantibodies against phospholipid compounds. Annexin V is used as a probe in the 'annexin V affinity assay' to detect cells that have expressed phosphatidylserine on the cell surface, a feature found in apoptosis and other forms of cell death.
Fusion-Tag: GST tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité