Human recombinant TNF alpha (soluble) (from E. coli)
Fournisseur: ProSci Inc.
90-222
PRSI90-222EA 400 EUR InStock
PRSI90-222
Human recombinant TNF alpha (soluble) (from E. coli)
Protéines et Peptides
Tumour necrosis factor (TNF, cachexin or cachectin and formerly known as tumour necrosis factor-alpha) is a cytokine involved in systemic inflammation and is a member of a group of cytokines that stimulate the acute phase reaction. The receptor TNF-R1 is activated by both the membrane-bound and soluble trimeric forms of TNF-alpha, whereas the receptor TNF-R2 only responds to the membrane-bound form of TNF-alpha (MultimericTNF-alpha (Prod. No. AG-40B-0019). Since the MultimericTNF-alpha mimics the membrane-bound form (forms oligomers higher than trimer), it is the only TNF-alpha protein that can activate the TNF-R2. For TNF-R1 activation, either "normal" TNF-alpha or MultimericTNF-alpha can be used."
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité