Human recombinant OX40L multimeric (from HEK293 cells)
Fournisseur: ProSci Inc.
90-239
PRSI90-239EA 567 EUR InStock
PRSI90-239
Human recombinant OX40L multimeric (from HEK293 cells)
Protéines et Peptides
OX40L [CD252; CD134L] is a membrane-expressed cytokine that belongs to the TNF ligand family. It acts as costimulator through interaction with its ligand OX40 on T cells, stimulating T cell activation, proliferation and cytokine production. OX40L is expressed on antigen presenting cells including B cells, dendritic cells, mast cells and endothelium. MultimericOX40L is a high activity construct in which two trimeric OX40L ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively activates T cell proliferation.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité