Human recombinant FN14 (from HEK293 cells)
Fournisseur: ProSci Inc.
90-242
PRSI90-242EA 528 EUR InStock
PRSI90-242
Human recombinant FN14 (from HEK293 cells)
Protéines et Peptides
TWEAK is a cytokine that controls proliferation, migration, differentiation, apoptosis, angiogenesis and inflammation. TWEAK binds to Fn14, a highly inducible cell-surface receptor that is linked to several intracellular signalling pathways, including the nuclear factor-B (NF-kappaB) pathway. The TWEAK–Fn14 axis plays a beneficial role in tissue repair following acute injury and may contribute to cancer, chronic autoimmune diseases and acute ischaemic stroke.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité